DuPont ™ Suprel ™ LS Coveralls
Offer a unique balance of breathability and barrier protection
$224.04 - $254.30
Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
19-165-213 Dupont Canada Inc
IC181SWH4X002500
Case of 25 Each for $254.30
19-165-211 Dupont Canada Inc
IC181SWH2X002500
Case of 25 Each for $224.04
Specifications
Description
Patented Suprel LS bi-component fabric offers breathability and barrier protection combined with soft feel and optimal comfort. Developed specifically for the life sciences, pharmaceutical, biotechnology and medical device manufacturing industries.
  • Softer feel and provides a greater ease of movement
  • Provides particulate and splash protection
  • Zipper-front closure, serged seams, white
  • Available sterilized or nonsterilized
Suprel LS is not flame-resistant. Should not be used around heat, flame, sparks, or in potential flammable or explosive environments.
Compliance
Meet ASTM™ standards for particle and bacterial filtration efficiency.