missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DuPont ™ Spunbonded Polypropylene Shoe Covers

Available with or without skid-resistant tread

$17.50 - $85.35

Specifications

Color Blue
Material Spunbond Polypropylene, Poly Film
For Use With (Application) Ideal for the challenges of dry and wet condition
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Size Sole Type Price Quantity  
Catalog Number Mfr. No. Size Sole Type Price Quantity  
191302717 Dupont Canada Inc
2854
X-Large Skid-resistant Case of 200 Each for $85.35
19-130-5321 Dupont Canada Inc
2850*
Universal With out skid-resistant Pack of 100 Each for $17.50
Case of 4 Pack for $64.84
19-130-2727 Dupont Canada Inc
2852
Universal Skid-resistant Case for $72.70
Specifications

Description

  • Made from nonwoven, spunbonded polypropylene fabric
  • 100% latex free
  • Color: Blue

Order Info

Packaged 100 per box, 4 boxes per case.

Description & Specifications

Specifications

Blue
Spunbond Polypropylene, Poly Film
Ideal for the challenges of dry and wet condition