DuPont™ ProShield™ NexGen™ Apron and Sleeves
Maximum protection at affordable cost
$88.76 - $180.57
Specifications
Color White
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Closure Type Seam Type Length (English) Material Price Quantity  
Catalog Number Mfr. No. Closure Type Seam Type Length (English) Material Price Quantity  
19-048-402A Dupont Canada Inc
NG273BWH00010000
Ties Bound 36 in. Tyvek™ Fabric Case of 100 Each for $180.57
29-634-224 Dupont Canada Inc
NG500SWH00020000
Zipper Serged 18 in. Fabric Case of 100 Pair for $88.76
Specifications
Description
DuPont ProShield NexGen is liquid resistant and provides a true aerosol barrier, with greater-than99% holdout of particles down to 0.35Glc mum. Proprietary microporous technology gives aprons and sleeves effective protective properties at an economical price. Perfect for applications involving nontoxic liquid or spray hazards.
DuPont ProShield NexGen is liquid resistant and provides a true aerosol barrier, with >99% holdout of particles down to 0.35μm. Proprietary microporous technology gives aprons and sleeves effective protective properties at an economical price. Perfect for applications involving nontoxic liquid or spray hazards.
Apron
  • Bib-style apron provides coverage of front and sides
  • With bound neck and ties
  • Length: 36 in. (91cm)
  • White
Sleeves
  • Elastic at both ends
  • Length: 18 in. (46cm)
  • White
Order Info
Aprons: Case of 100; Sleeves: Case of 200 (100 pairs).
Description & Specifications
Specifications
White