DuPont™ ProClean™ Limited-Use Cleanroom Frocks
Maximum containment short of a full coverage coverall. DuPont™ ProClean™ limited-use, full-length frocks offer the breathability and comfort of microporous film fabric.
Manufacturer:  DuPont™ PC270SWH4X00300B
 View more versions of this product
Catalog No. 29-635-5593
Description
Description
Proprietary polypropylene laminate fabric provides effective contamination control and protection from liquid-splash and dry-particulate cleanroom hazards.
  • White knee-length frocks
  • Microporous film fabric
  • Serged seams
  • Elastic wrist
  • Bound neck
  • Available with a five-snap or zipper front closure
  • Antistatic treated
Specifications
Specifications
Safety
White
4X-Large
46 to 52 in.
144.7cm
Snaps
Tunneled elastic
Serged
116.8 to 132 cm
57 in.
Documents