missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DuPont™ ProClean™ Bulk Packed Coveralls
Provide a barrier against particles and nonhazardous light liquid splash
Manufacturer: DuPont™ PC143SWH2X00250B
Description
- Lightweight
- Anti-static treated
- Low linting
- Made from a microporous composite fabric
- Serged seams
- Bound neck
- Elastic back
- Covered elastic wrist and ankles
- Zipper Closure
Specifications
Elastic | |
Fabric | |
White | |
Excellent barrier against particles and nonhazardous light liquid splash | |
185.92 to 198.12 cm | |
100.33 cm | |
Serged | |
43 to 46 in. | |
109.2 to 116.84 cm | |
Set |
Zipper | |
Elastic | |
Elastic | |
6.1 to 6.5 ft. | |
39.5 in. | |
Hooded | |
2X-Large | |
31.5 in. | |
80 cm | |
5.4 mil |