missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DuPont™ NexGen™ Protective Coveralls: With Hood and Boots

Affordable high-barrier protection

Manufacturer:  DuPont™ NG122SWHSM002500

 View more versions of this product

Catalog No. 29-634-232



Description

Description

Lightweight coveralls (1.8 oz. per sq. yd.) provide excellent barrier protection against nontoxic particle, liquid and spray hazards. True aerosol- and liquid-proof microporous barrier demonstrates >99% holdout of particles down to 0.35μm.

  • Breathable, cloth-like material treated to provide antistatic properties for use in controlled environments
  • Proprietary bilaminate composite fabric provides superior liquid penetration1
  • Compared favorably to leading fabric in physical property tests covering resistance to ripping and tearing2
  • Low linting for use in critical applications
  • Disposable coveralls can be easily discarded via landfill burial or proper incineration3
  • Long torso with polyester front zipper measures 26 in. long (66cm)
  • White

Compliance

Meets USDA, FDA, OSHA standards; ASTM™ D1777, ASTM D3776, ASTM D774, ASTM D5733, ASTM D5034, ASTM D257; AATCC 127 compliant.

1 Measured per ASTM™ F 903 in relation to the following substances: household bleach, diazinon (4 oz./gal.), hydraulic jack oil, machine oil, malathion (50%), motor oil (40W), sodium hydroxide and isocyanate-based paint hardener. Tests were performed by independent laboratories on material samples under laboratory conditions, not with actual garments or in actual onsite conditions.
2 ASTM D 751 (Grab Tensile Strength), ASTM D 1117 (Trapezoid Tear), ASTM D 3787 (Ball Burst).
3 Certified incinerable by independent laboratory. For details, contact your Fisher Safety Customer Service Representative at 1-800-772-6733.

Specifications

Specifications

Attached boots,Elastic
Fabric
White
Microporous Film, Laminated to a Spundbonded Polypropylene Substrate
Automotive refinishing, waste cleanup and remediation
1.52 to 1.73 m
78 cm
Serged
29.75 to 33.25 in.
75.56 to 84.45 cm
10 mil
Front zipper
USDA, FDA, OSHA, ASTM D1777, ASTM D3776, ASTM D774, ASTM D5733, ASTM D5034, ASTM D257, AATCC 127
Elastic
Yes
5 to 5.7 ft.
30.75 in.
Hooded
Small
26 in.
66.04 cm
SDS
Documents