Learn More
DuPont™ NexGen™ Protective Coveralls: With hood
Manufacturer: DuPont™ NG127SWH3X0025NP**
Description
Lightweight coveralls (1.8 oz. per sq. yd.) provide excellent barrier protection against nontoxic particle, liquid and spray hazards. True aerosol- and liquid-proof microporous barrier demonstrates >99% holdout of particles down to 0.35μm.
- Breathable, cloth-like material treated to provide antistatic properties for use in controlled environments
- Proprietary bilaminate composite fabric provides superior liquid penetration1
- Compared favorably to leading fabric in physical property tests covering resistance to ripping and tearing2
- Low linting for use in critical applications
- Disposable coveralls can be easily discarded via landfill burial or proper incineration3
- Long torso with polyester front zipper measures 26 in. long (66cm)
- White
1 Measured per ASTM™ F 903 in relation to the following substances: household bleach, diazinon (4 oz./gal.), hydraulic jack oil, machine oil, malathion (50%), motor oil (40W), sodium hydroxide and isocyanate-based paint hardener. Tests were performed by independent laboratories on material samples under laboratory conditions, not with actual garments or in actual onsite conditions.
2 ASTM D 751 (Grab Tensile Strength), ASTM D 1117 (Trapezoid Tear), ASTM D 3787 (Ball Burst).
3 Certified incinerable by independent laboratory. For details, contact your Fisher Safety Customer Service Representative at 1-800-772-6733.
Specifications
ASTM D751, D1117, D5034, D257, AATCC 127 | |
Serged | |
3X-Large | |
1.88 to 1.95m | |
37 in. | |
28.75 in. | |
10mil | |
Elastic | |
Yes | |
Fabric | |
47.75 to 51.25 in. |
Elastic | |
White | |
6.2 to 6.4 ft. | |
Automotive refinishing, waste cleanup and remediation | |
93.98cm | |
73.02cm | |
Zipper front | |
Hooded | |
Face opening, attached hood; Microporous Film, Laminated to a Spundbonded Polypropylene Substrate | |
121.28 to 130.17cm |