Please login to your online account to display your discounted pricing

DuPont™ NexGen™ Protective Coveralls with Elastic Wrists and Ankles

Affordable high-barrier protection

$151.25 - $188.49

Specifications

Ankle Style Elastic
Certifications/Compliance ASTM D751, D1117, D5034, D257, AATCC 127
Collar Type Lay down
Closure Type Zipper Front
Color White
View More Specs
Products
Catalog Number Mfr. No. Ankle Style Height (English) Cuff Style Size Price Quantity    

19065172

 
dupont canada inc
NG125SWH5X002500
Elastic 6.7 to 6.10 ft. Elastic 5X-Large (6 ft. 7 in. to 6 ft. 10 in.) Case of 25 Each for $188.49

29-634-239

 
dupont canada inc
NG125SWHSM002500
Elastic 5 to 5.7 ft. Elastic Small Case of 25 Each for $158.67

29-634-241

 
dupont canada inc
NG125SWHLG002500
Elastic 5.5 to 5.9 ft. Elastic Large Case of 25 Each for $151.25

29-634-242

 
dupont canada inc
NG125SWHXL002500
Elastic 5.8 to 6.2 ft. Elastic X-Large Case of 25 Each for $156.29

29-634-244

 
dupont canada inc
NG125SWH3X002500
Elastic 6.2 to 6.4 ft. Elastic 3X-Large Case of 25 Each for $165.16

29-634-240

 
dupont canada inc
NG125SWHMD002500
Elastic 5.3 to 5.7 ft. Elastic Medium Case of 25 Each for $156.29

29-634-245

 
dupont canada inc
NG125SWH4X002500
Elastic 6.4 to 6.7 ft. Elastic 4X-Large Case of 25 Each for $176.00

29-634-243

 
dupont canada inc
NG125SWH2X002500
Elastic 6 to 6.4 ft. Elastic 2X-Large Case of 25 Each for $151.84
Description & Specifications

Specifications

Ankle Style Elastic
Certifications/Compliance ASTM D751, D1117, D5034, D257, AATCC 127
Collar Type Lay down
Closure Type Zipper Front
Color White
Construction Microporous Film, Laminated to a Spundbonded Polypropylene Substrate
Cuff Style Elastic
Material Fabric
Seam Type Serged
Disposable Yes
For Use With (Application) Automotive refinishing, waste cleanup and remediation
Thickness 10mil

Lightweight coveralls (1.8 oz. per sq. yd.) provide excellent barrier protection against nontoxic particle, liquid and spray hazards. True aerosol- and liquid-proof microporous barrier demonstrates >99% holdout of particles down to 0.35μm.

  • Breathable, cloth-like material treated to provide antistatic properties for use in controlled environments
  • Proprietary bilaminate composite fabric provides superior liquid penetration1
  • Compared favorably to leading fabric in physical property tests covering resistance to ripping and tearing2
  • Low linting for use in critical applications
  • Disposable coveralls can be easily discarded via landfill burial or proper incineration3
  • Long torso with polyester front zipper measures 26 in. long (66cm)
  • White

1 Measured per ASTM™ F 903 in relation to the following substances: household bleach, diazinon (4 oz./gal.), hydraulic jack oil, machine oil, malathion (50%), motor oil (40W), sodium hydroxide and isocyanate-based paint hardener. Tests were performed by independent laboratories on material samples under laboratory conditions, not with actual garments or in actual onsite conditions.
2 ASTM D 751 (Grab Tensile Strength), ASTM D 1117 (Trapezoid Tear), ASTM D 3787 (Ball Burst).
3 Certified incinerable by independent laboratory. For details, contact your Fisher Safety Customer Service Representative at 1-800-772-6733.