missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DuPont™ NexGen™ Protective Coveralls: With Collar

Affordable high-barrier protection

Manufacturer:  DuPont™ NG120SWHSM002500

Catalog No. 29-634-225



Description

Description

Lightweight coveralls (1.8 oz. per sq. yd.) provide excellent barrier protection against nontoxic particle, liquid and spray hazards. True aerosol- and liquid-proof microporous barrier demonstrates >99% holdout of particles down to 0.35μm.

  • Breathable, cloth-like material treated to provide antistatic properties for use in controlled environments
  • Proprietary bilaminate composite fabric provides superior liquid penetration1
  • Compared favorably to leading fabric in physical property tests covering resistance to ripping and tearing2
  • Low linting for use in critical applications
  • Long torso with polyester front zipper measures 26 in. long (66cm)
  • White

1 Measured per ASTM™ F 903 in relation to the following substances: household bleach, diazinon (4 oz./gal.), hydraulic jack oil, machine oil, malathion (50%), motor oil (40W), sodium hydroxide and isocyanate-based paint hardener. Tests were performed by independent laboratories on material samples under laboratory conditions, not with actual garments or in actual onsite conditions.
2 ASTM D 751 (Grab Tensile Strength), ASTM D 1117 (Trapezoid Tear), ASTM D 3787 (Ball Burst).

Specifications

Specifications

Open
Fabric
Lay down
Open
Yes
5 to 5.7 ft.
31.25 in.
Collared
Small
26.5 in.
67.31 cm
Front zipper
ASTM D751, D1117, D5034, D257, AATCC 127
White
Microporous Film, Laminated to a Spundbonded Polypropylene Substrate
Automotive refinishing, waste cleanup and remediation
1.52 to 1.73 m
79.37 cm
Serged
29.75 to 33.25 in.
75.56 to 84.45 cm
10 mil
SDS
Documents
Product Certifications