missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DuPont™ NexGen™ Protective Coveralls: With Collar

Affordable high-barrier protection

Manufacturer:  DuPont™ NG120SWH4X002500

 View more versions of this product

Catalog No. 19-088-2143



Description

Description

Lightweight coveralls (1.8 oz. per sq. yd.) provide excellent barrier protection against nontoxic particle, liquid and spray hazards. True aerosol- and liquid-proof microporous barrier demonstrates >99% holdout of particles down to 0.35μm.

  • Breathable, cloth-like material treated to provide antistatic properties for use in controlled environments
  • Proprietary bilaminate composite fabric provides superior liquid penetration1
  • Compared favorably to leading fabric in physical property tests covering resistance to ripping and tearing2
  • Low linting for use in critical applications
  • Long torso with polyester front zipper measures 26 in. long (66cm)
  • White

1 Measured per ASTM™ F 903 in relation to the following substances: household bleach, diazinon (4 oz./gal.), hydraulic jack oil, machine oil, malathion (50%), motor oil (40W), sodium hydroxide and isocyanate-based paint hardener. Tests were performed by independent laboratories on material samples under laboratory conditions, not with actual garments or in actual onsite conditions.
2 ASTM D 751 (Grab Tensile Strength), ASTM D 1117 (Trapezoid Tear), ASTM D 3787 (Ball Burst).

Specifications

Specifications

Open
Fabric
Lay down
Open
Yes
6.4 to 6.7 ft.
37.5 in.
Collared
4X-Large
29.25 in.
74.29 cm
Front zipper
ASTM D751, D1117, D5034, D257, AATCC 127
White
Microporous Film, Laminated to a Spundbonded Polypropylene Substrate
Automotive refinishing, waste cleanup and remediation
1.95 to 2.04 m
95.25 cm
Serged
51.25 to 54.75 in.
130.17 to 139.06 cm
10 mil
SDS
Documents