Please login to your online account to display your discounted pricing

DuPont™ Limited-Use Coated Tyvek ™ Apparel, Saranex-23P-Coated Tyvek Coveralls with Hood, Elasticized Wrists and Ankles, Bound Seams

Offers an inherent barrier to small size particles

$294.70 - $315.16

Specifications

Ankle Style Elastic
Certifications/Compliance ASTM D1777, D3776, D3786, D5034
Closure Type Zipper, Storm Flap
Color White
Construction Saranex™ 23-P Film Laminated to Tyvek™ Fabric
View More Specs
Products
Catalog Number Mfr. No. Height (Metric) Length (Metric) Sleeve Size Size (Metric) Chest Price Quantity    

29-801-439

 
dupont canada inc
SL127BWH2X001200
1.82 to 1.95m 97.15cm 2X-Large 112.39 to 121.28cm Case of 12 Each for $315.16

29-801-438

 
dupont canada inc
SL127BWHXL001200
1.76 to 1.88m 92.71cm X-Large 104.77 to 113.66cm Case of 12 Each for $294.70
Description & Specifications

Specifications

Ankle Style Elastic
Certifications/Compliance ASTM D1777, D3776, D3786, D5034
Closure Type Zipper, Storm Flap
Color White
Construction Saranex™ 23-P Film Laminated to Tyvek™ Fabric
Cuff Style Elastic
Includes Elastic around the face, elastic wrist
Material Tyvek™ Fabric
Seam Type Bound
Disposable Yes
For Use With (Application) Chemical mixing, hazardous remediation, emergency medical response, paint spraying and nuclear environments
Hood Type Attached
Size (Metric) Waist 142.24cm
Thickness 0.33mm
Weight (English) 0.22 lb.
Weight (Metric) 0.1kg

Applications include: lead and asbestos abatement/remedation, general maintenance/operations, spray painting, general clean-up

  • Comfort fit design based on extensive wearer input
  • Designed to cover neck and chin and fit around respirator face mask
  • No films or laminates to abrade or wear away
  • Elastic opening for tighter fit at wrist, ankle