missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DuPont™ Tyvek™ IsoClean™ Series 182B Coveralls, Bulk
Tyvek IsoClean delivers an ideal balance of protection, durability and comfort. Bulk packaged in double transparent poly liners.
Manufacturer: DuPont™ IC182BWH4X002500
Description
Made using a patented flash spinning process, Tyvek provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash.- Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
- Coverall has elastic openings for tighter fit at wrist and ankle
- Bound neck for lower particle shedding
- Front zipper closure for easy donning and doffing
- Raglan sleeve design for greater range of motion and comfort
- White
Specifications
Elastic | |
Tyvek™ IsoClean™ Flashspun Polyethylene | |
Elastic | |
6.3 to 7 ft. | |
42 in. | |
Bound | |
52 to 58 in. | |
132.08 to 147.32 cm | |
Raglan |
Zipper | |
White | |
Premium protection for controlled environments and excellent inherent barrier to dry particles, microorganisms and non-hazardous liquids | |
192.02 to 213.3 cm | |
106.68 cm | |
4X-Large | |
35 in. | |
88.9 cm | |
5 mil |