DuPont™ Tyvek™ IsoClean™ Series 182B Coveralls, Bulk
Tyvek IsoClean delivers an ideal balance of protection, durability and comfort. Bulk packaged in double transparent poly liners.
Manufacturer:  DuPont™ IC182BWH3X002500
 View more versions of this product
Catalog No. 19-088-1120F
Description
Description
Made using a patented flash spinning process, Tyvek provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash.
  • Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
  • Coverall has elastic openings for tighter fit at wrist and ankle
  • Bound neck for lower particle shedding
  • Front zipper closure for easy donning and doffing
  • Raglan sleeve design for greater range of motion and comfort
  • White
Specifications
Specifications
Elastic
Tyvek™ IsoClean™ 182 Series flashspun polyethylene
White
Premium protection for controlled environments and excellent inherent barrier to dry particles, microorganisms and non-hazardous liquids
Bound
Raglan
Zipper
Bound
Elastic
Collared
3X-Large
5 mil