DuPont™ Tyvek™ IsoClean™ Series 182B Coveralls, Bulk
Tyvek IsoClean delivers an ideal balance of protection, durability and comfort. Bulk packaged in double transparent poly liners.
Manufacturer:  DuPont™ IC182BWHMD002500
 View more versions of this product
Catalog No. 19-088-1120B
Description
Description
Made using a patented flash spinning process, Tyvek provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash.
  • Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
  • Coverall has elastic openings for tighter fit at wrist and ankle
  • Bound neck for lower particle shedding
  • Front zipper closure for easy donning and doffing
  • Raglan sleeve design for greater range of motion and comfort
  • White
Specifications
Specifications
Elastic
Tyvek™ IsoClean™ Flashspun Polyethylene
White
Premium protection for controlled environments and excellent inherent barrier to dry particles, microorganisms and non-hazardous liquids
167.64 to 155.44 cm
93.98 cm
Bound
36 to 38 in.
91.44 to 96.52 cm
Raglan
Zipper
Bound
Elastic
5.5 to 5.10 ft.
37 in.
Collared
Medium
32.5 in.
82.55 cm
5 mil