missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DuPont™ Tyvek™ IsoClean™ Series 182B Coveralls, Clean-Processed and Sterile
Cleanroom coveralls deliver the best balance of protection, comfort, and durability. Clean-processed and gamma sterilized to SAL of 10-6.
Supplier: DuPont™ IC182BWH3X0025CS
Description
Made using a patented flash spinning process, Tyvek provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash.- Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
- Coverall has elastic openings for tighter fit at wrist and ankle
- Bound neck for lower particle shedding
- Front zipper closure for easy donning and doffing
- Raglan sleeve design for greater range of motion and comfort
- White
Specifications
Cleanroom Coveralls | |
Tyvek™ | |
Elastic | |
Zipper | |
Sterile | |
IC182BWH | |
35 in. | |
88.9 cm | |
5.4 mil | |
41 in. | |
6.2 to 6.6 ft. |
3X-Large | |
White | |
Elastic | |
Bound | |
Clean and Sterile | |
46 to 52 in. | |
116.8 to 132.08 cm | |
Raglan | |
Premium protection for controlled environments and inherent barrier to dry particles, microorganisms, and non-hazardous liquids | |
104.14 cm | |
188.97 to 201.16 cm |