DuPont™ Tyvek™ IsoClean™ Series 182B Coveralls, Clean-Processed and Sterile
DuPont Tyvek Cleanroom Coveralls delivers the best balance of protection, comfort and durability. Clean-processed and gamma sterilized to SAL of 10-6
Manufacturer:  DuPont™ IC182BWH2X0025CS
 View more versions of this product
Catalog No. 19-088-1123E
Description
Description
Made using a patented flash spinning process, Tyvek provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash.
  • Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
  • Coverall has elastic openings for tighter fit at wrist and ankle
  • Bound neck for lower particle shedding
  • Front zipper closure for easy donning and doffing
  • Raglan sleeve design for greater range of motion and comfort
  • White
Specifications
Specifications
Elastic
Tyvek™ IsoClean™ Flashspun Polyethylene
White
Clean and Sterile
6.1 to 6.5 ft.
40 in.
Bound
43 to 46 in.
109.2 to 116.84 cm
Raglan
Zipper
Sterile
Elastic
Premium protection for controlled environments and excellent inherent barrier to dry particles, microorganisms and non-hazardous liquids
185.92 to 198.12 cm
101.6 cm
2X-Large
35 in.
88.9 cm
5.4 mil