DuPont™ Tyvek™ IsoClean™ Series 182B Coveralls, Clean-Processed and Sterile
DuPont Tyvek Cleanroom Coveralls delivers the best balance of protection, comfort and durability. Clean-processed and gamma sterilized to SAL of 10-6
Manufacturer:  DuPont™ IC182BWHLG0025CS
 View more versions of this product
Catalog No. 19-088-1123C
Description
Description
Made using a patented flash spinning process, Tyvek provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash.
  • Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
  • Coverall has elastic openings for tighter fit at wrist and ankle
  • Bound neck for lower particle shedding
  • Front zipper closure for easy donning and doffing
  • Raglan sleeve design for greater range of motion and comfort
  • White
Specifications
Specifications
Elastic
Tyvek™ IsoClean™ Flashspun Polyethylene
Bound
Elastic
Premium protection for controlled environments and excellent inherent barrier to dry particles, microorganisms and non-hazardous liquids
176.78 to 182.88 cm
96.52 cm
Bound
38 to 40 in.
96.52 to 101.6 cm
Raglan
Zipper
Sterile
White
Clean and Sterile
5.8 to 6 ft.
38 in.
Collared
Large
33.5 in.
85.09 cm
5.4 mil