missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DuPont™ Tyvek™ IsoClean™ Series 182B Coveralls, Clean-Processed and Sterile
DuPont Tyvek Cleanroom Coveralls delivers the best balance of protection, comfort and durability. Clean-processed and gamma sterilized to SAL of 10-6
Manufacturer: DuPont™ IC182BWHMD0025CS
Description
Made using a patented flash spinning process, Tyvek provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash.- Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
- Coverall has elastic openings for tighter fit at wrist and ankle
- Bound neck for lower particle shedding
- Front zipper closure for easy donning and doffing
- Raglan sleeve design for greater range of motion and comfort
- White
Specifications
Elastic | |
Tyvek™ IsoClean™ Flashspun Polyethylene | |
Bound | |
Elastic | |
Premium protection for controlled environments and excellent inherent barrier to dry particles, microorganisms and non-hazardous liquids | |
167.64 to 155.44 cm | |
93.98 cm | |
Bound | |
36 to 38 in. | |
91.44 to 96.52 cm | |
Raglan |
Zipper | |
Sterile | |
White | |
Clean and Sterile | |
5.5 to 5.10 ft. | |
37 in. | |
Collared | |
Medium | |
32.5 in. | |
82.55 cm | |
5.4 mil |