DuPont ™ Fully Encapsulated Level A Tychem ™ TK ™ Suits, NFPA 1991
Excellent chemical holdout for use in hazmat situations
$3,822.40 - $4,059.64
Specifications
Includes Expanded faceshield, 4 exhaust valves, internal adjustment waist belt, attached internal North SSG - Silver Shield™/4H™/external Neoprene size 10 gloves, separate Kevlar™ knit over gloves, socks with outer boot flaps, soles, separate Aluminize
Material Tychem™ TK
Seam Type Double taped
For Use With (Application) Industrial, HazMat and domestic preparedness applications
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Style Color Size Price Quantity  
Catalog Number Mfr. No. Style Color Size Price Quantity  
29-635-0067 Dupont Canada Inc
TK600TLYXL000100
Front entry Silver X-Large Case for $3,822.40
19-161-729 Dupont Canada Inc
TK601TLYXL000100
Rear entry Silver X-Large Each for $3,829.86
19-161-730 Dupont Canada Inc
TK601TLY2X000100
Rear entry Silver 2X-Large Each for $4,059.64
29-635-0068 Dupont Canada Inc
TK600TLY2X000100
Front entry Silver 2X-Large Case for $4,051.72
Specifications
Description
Made from a high-strength nonwoven with nonhalogenated films on either side.
Inner Suit
  • Double-taped seams
  • Three-layer EX faceshield (PVC 40mil/PTFE 5mil/PVC 20mil)
  • Gastight zipper closure
  • Double-storm flap
  • Attached three-layer gloves (Barrier™/neoprene/Kevlar™ knit) and sock boots
Outer Suit
  • Aluminized fiberglass with sewn NOMEX™ seams
  • EX faceshield (Lexan™ 20mil)
  • Hook-and-loop closure
  • Storm flap
  • Hemmed wrists
Description & Specifications
Specifications
Expanded faceshield, 4 exhaust valves, internal adjustment waist belt, attached internal North SSG - Silver Shield™/4H™/external Neoprene size 10 gloves, separate Kevlar™ knit over gloves, socks with outer boot flaps, soles, separate Aluminize
Tychem™ TK
Double taped
Industrial, HazMat and domestic preparedness applications