missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DuPont™ DuraTrac™ Shoe Covers
Skid-resistant shoe covers provide an excellent barrier to dry particles and liquids
$170.17 - $175.44
Specifications
Coating | Polyethylene |
---|---|
Closure Type | Elastic |
Color | Blue |
Material | Polypropylene |
Product Type Specs | Shoe Covers |
Alert:
Due to extraordinary demand patterns, this item may experience longer lead times and shipping/delivery dates may not be accurate.
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
17-988-016
|
DuPont™
DT440SBULG03000B |
Large |
Case of 300 Each for $175.44
|
|
17988017
|
DuPont™
DT440SBUXL03000B |
X-Large |
Case of 300 Each for $170.17
|
|
Description
- Spunbonded polypropylene with polyethylene extrusion coating
- Serged seams
- Elastic opening with defined heel and toe
- Bulk packed double poly liners
- Skid-resistant surface helps prevent accidents in wet or dry conditions
Specifications
Polyethylene | |
Blue | |
Shoe Covers | |
Ideal to provide an excellent barrier to dry particles and liquids and helps prevent accidents in wet or dry conditions | |
Serged, heat sealed |
Elastic | |
Polypropylene | |
Non-skid | |
Yes |