DuPont ™ Cool-Guard ™ Personal Cooling Vest
State-of-the-art cooling technology maintains proper body temperature
$360.65 - $360.65
Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With (Application) Quantity Product Type Price Quantity  
Catalog Number Mfr. No. For Use With (Application) Quantity Product Type Price Quantity  
29-663-456 Dupont Canada Inc
996000BU00000100
Aids in reducing potential for heat stress while wearing chemical protective garments Each Cool-Guard™ Vest Each for $360.65
Specifications
Description
The Cool-Guard vest provides cooling without the discomfort, vasoconstriction, or moisture buildup of typical icepack systems.
  • Phase-change gel packs absorb heat generated by the body
  • Provide 1.5 to 2 hours of cooling at 43.3°C (110°F)
  • Gel packs recharge quickly in ice water within 20 minutes
  • Ergonomically designed blue Banox™ vest is light and flexible
  • Fitted with adjustable straps so one size fits all
  • Flame-retardant
Order Info
Total weight (including phase-change packs): 3.88 lb. (1.8kg)