missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dulcitol, 99%

Useful reagent in bacteriology

$252.91 - $351.61

Chemical Identifiers

CAS 608-66-2
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.172
MDL Number MFCD00064288
InChI Key FBPFZTCFMRRESA-GUCUJZIJSA-N
Synonym dulcitol, galactitol, dulcite, d-galactitol, dulcose, euonymit, melampyrin, melampyrit, d-dulcitol, 2r,3s,4r,5s-hexane-1,2,3,4,5,6-hexaol
PubChem CID 11850
ChEBI CHEBI:16813
IUPAC Name (2R,3S,4R,5S)-hexane-1,2,3,4,5,6-hexol
SMILES C(C(C(C(C(CO)O)O)O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6320630 View Documents Alfa Aesar™
J6320630
250g Each for $252.91
 
AAJ6320636 View Documents Alfa Aesar™
J6320636
500g Each for $351.61
 
Specifications

Chemical Identifiers

608-66-2
182.172
FBPFZTCFMRRESA-GUCUJZIJSA-N
11850
(2R,3S,4R,5S)-hexane-1,2,3,4,5,6-hexol
C6H14O6
MFCD00064288
dulcitol, galactitol, dulcite, d-galactitol, dulcose, euonymit, melampyrin, melampyrit, d-dulcitol, 2r,3s,4r,5s-hexane-1,2,3,4,5,6-hexaol
CHEBI:16813
C(C(C(C(C(CO)O)O)O)O)O

Specifications

1.47
275°C to 280°C (1mmHg)
187°C to 191°C
608-66-2
MFCD00064288
14,4332
FBPFZTCFMRRESA-GUCUJZIJSA-N
(2R,3S,4R,5S)-hexane-1,2,3,4,5,6-hexol
11850
182.17
99%
Hygroscopic
White
250g
C6H14O6
1721903
dulcitol, galactitol, dulcite, d-galactitol, dulcose, euonymit, melampyrin, melampyrit, d-dulcitol, 2r,3s,4r,5s-hexane-1,2,3,4,5,6-hexaol
C(C(C(C(C(CO)O)O)O)O)O
182.172
CHEBI:16813
Powder
Dulcitol
Safety and Handling

Safety and Handling

TSCA : TSCA