Please login to your online account to display your discounted pricing

Dulbecco Modification of Eagle's Med., 1X Liq., w/o L-Glut., L-Cyst., L-Meth., L-Cysteine, MP Biomedicals

$107.82 - $107.82

Specifications

pH 6.9 to 7.5
Solution Type DMEM
Storage 2° to 8°C
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN1642454

 
mp biomedicals
1642454
500mL Each for $107.82
Description & Specifications

Specifications

pH 6.9 to 7.5
Solution Type DMEM
Storage 2° to 8°C