Please login to your online account to display your discounted pricing

DSP-4, Tocris

Adrenergic neurotoxin

$113.57 - $478.51

Chemical Identifiers

Chemical Name or Material DSP-4
Assay Percent Range >99%
Molecular Formula C11H15BrClN·HCl
Formula Weight 313.06
Synonym N-(2-Chloroethyl)-N-ethyl-2-bromobenzylamine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

295810

 
r & d systems
RDS295810
10mg Each for $113.57

295850

 
r & d systems
RDS295850
50mg Each for $478.51
Description & Specifications

Specifications

Formula Weight 313.06
Quantity 10mg
Molecular Formula C11H15BrClN·HCl
CAS 40616-75-9
Chemical Name or Material DSP-4
Assay Percent Range >99%
Recommended Storage Desiccate at Room Temperature
Synonym N-(2-Chloroethyl)-N-ethyl-2-bromobenzylamine hydrochloride
Safety and Handling

Desiccate at Room Temperature