Please login to your online account to display your discounted pricing
Drummond Scientific Fixed Volume Microdispensers Replacement Parts Kits
Manufacturer: drummond scientific  3000325K
 View more versions of this product
Catalog No. DRM3000325K
  • / Each
Description & Specifications
Specifications
For Use With (Equipment) 25μL Drummond™ fixed volume microdispenser
Includes 1 Plunger, 2 O-rings, 1 collet, 1 coil spring, 3 PTFE tip, 1 spring guide
Product Type Microdispenser Replacement Parts Kit