Drummond™ Dialamatic Microdispensers
Permit accurate fractional deliveries
$826.21 - $826.21
Specifications
Accuracy ±1%
Number of Channels 1
Type Dialamatic Microdispenser
For Use With (Application) Adjustable, direct reading plunger sets volume
Includes 50 Borosilicate Glass bores with Stainless Steel plunger
View More Specs
Includes: 100μL size comes with 50 glass bores; all others with 100 glass bores.
Products 4
Catalog Number Mfr. No. Volume (Metric) Increments Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Increments Price Quantity  
21-170-15B
Accessories View Documents Promotion Details
Drummond™
3000225
25 μL 1μL
Each for $826.21
21-170-15C
Accessories View Documents Promotion Details
Drummond™
3000250
50 μL 2μL
Each for $826.21
21-170-15A
Accessories View Documents Promotion Details
Drummond™
3000210
10 μL 1μL
Each for $826.21
21-170-15D
Accessories View Documents Promotion Details
Drummond™
3000275
100 μL 10μL
Each for $826.21
Description
Description
Four adjustable models cover volume range 1 to 100μL. Threaded plunger is graduated for accurate deliveries. Turn plunger to desired volume.
  • Two scales on 10 and 100μL sizes simplify volume selection
  • Accuracy to ±1% at full volume
  • Aluminum construction
  • Borosilicate glass bores with stainless steel plungers; PTFE tips on plungers 25μL or larger
Specifications
Specifications
±1%
Dialamatic Microdispenser
50 Borosilicate Glass bores with Stainless Steel plunger
1
Adjustable, direct reading plunger sets volume