Please login to your online account to display your discounted pricing
Drum Hygrothermograph
Economical and easy to use.
$48.95 - $514.65
Specifications
For Use With (Application) Measuring and recording both humidity and temperature
View More Specs
Includes:  55 Sheets of seven-day, ∞C chart paper and one AA battery.
Products
Catalog Number Mfr. No. Includes Product Type Price Quantity    
S65279
fisher science education
WD-35701-00
1 AA battery, Paper, chart pens, and replacement battery, built-in LCD clock, 55 sheets of seven day, chart Paper Drum hygrothermograph Each for $514.65
S65378
fisher science education
WD-09368-80
55 Sheets of seven-day, chart paper and one AA battery Red felt-tip pen Pack of 6 Each for $48.95
Description & Specifications
Specifications
For Use With (Application) Measuring and recording both humidity and temperature
  • Uses a humidity-sensing coil for accurate, consistent readings
  • Built-in LCD clock in base lets you view time at a glance
  • Drive is quartz-controlled for accuracy and operates for six months on one AA battery
  • Dimensions: 4.25L x 4.875W x 7.25 in.H (10.79 x 12.38 x 18.41cm)
  • Shipping weight: 3 lb.
Order pens separately.