Please login to your online account to display your discounted pricing
Drilled/Assorted Balls Set
Can be used for all pendulum experiments.
Manufacturer: fisher science education  DBLST12
 View more versions of this product
Catalog No. S07037
  • / Each
Description & Specifications
Specifications
For Use With (Application) Conducting pendulum experiment
Includes 3mm holes, Two each of 25mm dia. balls made of Brass, Aluminum, Steel, Wood, cork and Copper
Product Type Drilled Ball Set