United Scientific Supplies Drilled/Assorted Balls Set
Can be used for all pendulum experiments.
$109.30 - $109.30
Specifications
For Use With (Application) Conducting pendulum experiment
View More Specs
Includes: Drilled Ball Set: Two each of 25mm diameter balls made of brass, aluminum, steel, wood, cork and copper
Assorted Ball Set: 25mm diameter drilled balls of brass, aluminum, wood, steel, copper and lead; two 25mm solid steel balls (one plain and one with bifilar suspension lugs); 38mm balls of brass, steel and lead with hooks; and one 25mm solid cork ball
Products 1
Catalog Number Mfr. No. Product Type Includes Price Quantity  
Catalog Number Mfr. No. Product Type Includes Price Quantity  
S07037 View Documents United Scientific Supplies
DBLST12
Drilled Ball Set 3mm holes, Two each of 25mm dia. balls made of Brass, Aluminum, Steel, Wood, cork and Copper Each for $109.30
Description
Description
  • Balls made of different materials and sizes
  • Drilled balls have 3mm holes in center
  • Packed in plastic storage box
Specifications
Specifications
Conducting pendulum experiment