Please login to your online account to display your discounted pricing

Drierite™, without indicator, 6 mesh, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: fisher scientific acros  219045000

 View more versions of this product

Catalog No. AC219045000

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 136.14
Quantity 500g
Physical Form Granular
Chemical Name or Material Drierite™
Packaging Glass bottle
CAS 7778-18-9
Composition Typical chemical analysis: CaO: 40.80%, SO3: 58.80%, CO2: 0.60%, R2O: 0.14%, SiO2 and insolubles: 0.15%, NaCl: 0.18%
Molecular Formula CaO4S
Health Hazard 1

Specifications

Chemical Name or Material Drierite™
CAS 7778-18-9
Molecular Formula CaO4S
Formula Weight 136.14
Accessories

SDS & Safety Documents