missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Drierite™, without indicator, 6 mesh, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 219040020
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent, IndicatorSpecifications
1450.0°C | |
Drierite™ | |
2 kg | |
MFCD00010912 | |
Solubility in water: ca. 0.25g/L (20°C). Other solubilities: insoluble in organic liquids, slightly soluble in glycerol | |
[Ca++].[O-]S([O-])(=O)=O | |
136.13 | |
CHEBI:31346 |
Granular | |
7778-18-9 | |
CaO4S | |
Glass Bottle | |
OSGAYBCDTDRGGQ-UHFFFAOYSA-L | |
calcium sulfate | |
24497 | |
136.14 |
Chemical Identifiers
7778-18-9 | |
136.13 | |
OSGAYBCDTDRGGQ-UHFFFAOYSA-L | |
CHEBI:31346 |
CaO4S | |
MFCD00010912 | |
24497 | |
[Ca++].[O-]S([O-])(=O)=O |
Safety and Handling
RUO – Research Use Only