missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Drierite™, without indicator, 4 mesh, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 219030100
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Adsorbent, IndicatorSpecifications
Drierite™ | |
10kg | |
Plastic drum | |
OSGAYBCDTDRGGQ-UHFFFAOYSA-L | |
calcium;sulfate | |
24497 | |
136.14 |
7778-18-9 | |
CaO4S | |
Solubility in water: ca. 0.25g/L (20°C). Other solubilities: insoluble in organic liquids, slightly soluble in glycerol | |
[O-]S(=O)(=O)[O-].[Ca+2] | |
136.134 | |
CHEBI:31346 |
Safety and Handling
RUO â Research Use Only