Learn More
Drierite™, regular (CaSO{4}), Lab Grade
CAS: 7778-18-9 | CaO4S | 136.13 g/mol
Supplier: Thermo Scientific Chemicals 043056P5
Description
Drierite™, indicating (CaSO{4}), Lab Grade is used as a desiccant. It is used as a drying agent utilized for the efficient and rapid drying of air, industrial gases, refrigerants, organic liquids and solids. Further, it is used to maintain a dry atmosphere in storage spaces, vaults, commercial packages, and other enclosures. In addition to this, it is involved in the protection of hygroscopic materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Granular | |
| Drierite | |
| 2.96g/cm3 | |
| CaO4S | |
| Odorless | |
| OSGAYBCDTDRGGQ-UHFFFAOYSA-L | |
| Hygroscopic | |
| 136.13 | |
| CHEBI:31346 |
| White | |
| 7778-18-9 | |
| 5 lb. | |
| MFCD00010912 | |
| Soluble in water. | |
| [Ca++].[O-]S([O-])(=O)=O | |
| calcium sulfate | |
| 24497 | |
| Laboratory |
Chemical Identifiers
| 7778-18-9 | |
| 136.13 | |
| OSGAYBCDTDRGGQ-UHFFFAOYSA-L | |
| CHEBI:31346 | |
| [Ca++].[O-]S([O-])(=O)=O |
| CaO4S | |
| MFCD00010912 | |
| 24497 | |
| calcium sulfate |
Safety and Handling
H315-H319: Causes skin irritation. Causes serious eye irritation.
EINECSNumber : 231-900-3
RTECSNumber : WS6920000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only