Learn More
Drierite™, with indicator, 10-20 mesh, Thermo Scientific Chemicals
$119.79 - $1354.82
Chemical Identifiers
CAS | 7778-18-9 |
---|---|
Molecular Formula | CaO4S |
Molecular Weight (g/mol) | 136.13 |
MDL Number | MFCD00010912 |
InChI Key | OSGAYBCDTDRGGQ-UHFFFAOYSA-L |
PubChem CID | 24497 |
ChEBI | CHEBI:31346 |
SMILES | [Ca++].[O-]S([O-])(=O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC350015000
|
Thermo Scientific Chemicals
350015000 |
500 g | Glass Bottle |
Each for $119.79
|
|
||||
AC350010020
|
Thermo Scientific Chemicals
350010020 |
2 kg | Glass Bottle |
Each for $378.87
|
|
||||
AC350010100
|
Thermo Scientific Chemicals
350010100 |
10 kg | Plastic Drum |
Each for $1,354.82
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent, IndicatorChemical Identifiers
7778-18-9 | |
136.13 | |
OSGAYBCDTDRGGQ-UHFFFAOYSA-L | |
CHEBI:31346 |
CaO4S | |
MFCD00010912 | |
24497 | |
[Ca++].[O-]S([O-])(=O)=O |
Specifications
1450.0°C | |
Blue | |
500 g | |
MFCD00010912 | |
Glass Bottle | |
0.29% | |
[Ca++].[O-]S([O-])(=O)=O | |
136.13 | |
CHEBI:31346 | |
Drierite™ |
Irregular Granules | |
7778-18-9,7646-79-9 | |
CaO4S | |
3077 | |
Solubility in water: slightly soluble. Other solubilities: slightly soluble in glycerol, practically insoluble in most common organic, solvents. SiO2 and insolubles: 0.58% | |
OSGAYBCDTDRGGQ-UHFFFAOYSA-L | |
calcium sulfate | |
24497 | |
136.13 |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Suspected of causing genetic defects. May cause cancer by inhalation. May damage fertility. Causes serious eye irritation. Toxic to aquatic life with long lasting effects.
GHS P Statement
Obtain special instructions before use. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing. Wear protective gloves/protective clothing/eye protection/face protection. If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician. IF exposed or concerned: Get medical advice/attention.
Danger
RUO – Research Use Only