Learn More
Thermo Scientific™ DreamTaq DNA Polymerase (5 U/μL)
Get higher sensitivity, longer PCR products and higher yields in all standard PCR applications with this enhanced Taq DNA polymerase.
Supplier: Thermo Scientific™ EP0705
Description
DreamTaq DNA Polymerase is an enhanced Taq DNA polymerase optimized for all standard PCR applications. It ensures higher sensitivity, longer PCR products and higher yields compared to conventional Taq DNA polymerase. DreamTaq DNA Polymerase uses the same reaction set-up and cycling conditions as conventional Taq DNA polymerase. Extensive optimization of reaction conditions is not required. It is supplied with optimized DreamTaq buffer, which includes 20Â mM MgCl2.
DreamTaq PCR Master Mix (2X) is a ready-to-use solution containing DreamTaq DNA Polymerase, optimized DreamTaq buffer, MgCl2, and dNTPs. This pre-mixed formulation saves time and reduces contamination due to the fewer pipetting steps required for PCR set up. The master mix retains all features of DreamTaq DNA Polymerase. It is capable of robust amplification of up to 6 kb from genomic DNA and up to 20 kb from viral DNA.
Highlights
- Robust amplification with minimal optimization
- High yields of PCR products
- Higher sensitivity compared to conventional Taq DNA polymerase
- Amplification of long targets up to 6Â kb from genomic DNA and up to 20Â kb from viral DNA
- Incorporates modified nucleotides, but does not incorporate dUTP
Specifications
Contains: • 10 x 100 μL DreamTaq DNA Polymerase (5 U/μL) • 20 x 1.25 mL 10X DreamTaq Buffer (includes 20 mM MgCl2) Store at -20°C. |
|
DreamTaq DNA Polymerase | |
DreamTaq™ | |
1 X | |
3'-A |
Low | |
Standard | |
4000 x 250 ReactionsμL | |
No | |
Standalone |
For Research Use Only. Not for use in diagnostic procedures.