Please login to your online account to display your discounted pricing

Thermo Scientific DreamTaq Hot Start PCR Master Mix


Thermo Scientific DreamTaq Hot Start DNA Polymerase is the product of choice when you need consistently robust amplification with minimal optimization steps

Manufacturer: thermo scientific  K9011

 View more versions of this product

DreamTaq Hot Start PCR Master Mix, 2X; water, nuclease-free

Catalog No. FERK9011

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Routine PCR, Hot Start PCR, Genotyping, colony PCR, RT-PCR
Format 2X Master Mix
Storage Requirements Store at -20C.
Includes DreamTaq Hot Start PCR Master Mix, 2X; water, nuclease-free
Overhang 3′-A
Quantity 200 rxns

Thermo Scientific™ DreamTaq™ Hot Start DNA Polymerase is an enhanced hot start Taq DNA polymerase that ensures higher PCR specificity, sensitivity and yields compared to conventional hot start Taq DNA polymerases. DreamTaq Hot Start DNA Polymerase uses antibody based inhibition of DNA polymerase activity at ambient temperatures, thus preventing the amplification of non-specific products prior to the amplification step.

Better reaction outcomes: 
  • Higher yield of target amplicons from low template amounts.
  • Increased reaction specificity due to reliable hot-start technology.
  • Wider range of amplicon lengths - routine amplification of genomic DNA fragments up to 6kb.
Enhanced convenience:
  • Room temperature reaction set-up.
  • Minimized optimization of Mg2+ concentration and of primer annealing temperatures due to optimized reaction buffer.  
  • Simplified workflow and minimized pipetting steps with 2X master mix format.  
Amplicon size (complex DNA): 6 kb
Fidelity (vs. Taq): 1X

Store at -20C.