Please login to your online account to display your discounted pricing
Dräger™ Short-Term Detector Tubes: Alcohol
Detector tubes
$116.93 - $140.79
Specifications
Description Detector Tubes
Quantity 10
View More Specs
Products
Catalog Number Mfr. No. Humidity Range Measurement Range Tube Type Gases Measured Price Quantity    
18-565
dräger™
8101631
15mg/L 25 to 5000 (Methanol), 50 to 4000 (i-Propanol) Alcohol 25/a Alcohol Pack of 10 Each for $140.79
17-985-176
dräger™
CH29701
Less than20mg/L 100 to 3000 Alcohol 100/a Alcohol Pack of 10 Each for $116.93
Description & Specifications
Specifications
Description Detector Tubes
Quantity 10
Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.
Note: Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.