missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DR3 Mouse anti-Human, PE, Clone: JD3, eBioscience™

Mouse Monoclonal Antibody

$157.08 - $368.28

Specifications

Antigen DR3
Clone JD3
Host Species Murine
Gene Alias TRAMP, Apo-3, TRAIL Receptor family, Tyrosine-rich acidic matrix protein
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5011091 Life Technologies
LS12660342
100 Tests Each for $368.28
5011092 Life Technologies
LS12660341
25 Tests Each for $157.08
Specifications

Description

The JD3 monoclonal antibody reacts with human DR3, also known as Apo-3, WSL-1, TRAMP, LARD, DDR3, and TR3. DR3, a novel death receptor is expressed at low level by monocytes and granulocytes. Interaction of DR3 with its ligand activates NF-kappa B pathway and induces apoptosis. Initially, DR3 was thought to be a receptor for TWEAK, but further studies have shown that TWEAK could induce apoptosis via receptors distinct from DR3. JD3 has been reported for use in flow cytometric analysis.
Description & Specifications

Specifications

DR3
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
JD3
TRAMP, Apo-3, TRAIL Receptor family, Tyrosine-rich acidic matrix protein
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.125μg)/test