missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DR Instruments Biology Dissecting Kit

Avaliable in single fold vinyl case.

Manufacturer:  DR Instruments 77

Catalog No. S06836




Includes:
  • Student dissecting scissors 4.5 in.
  • Dissecting student forceps 4.5 in., chrome-plated
  • Dissecting scalpel
  • Two dissecting teasing needles (one straight and one angular) with metal chuck
  • Dissecting mall probe, chrome, 6 in.
  • Glass dropper
  • Ruler 6 in.
  • Single fold vinyl case

Description

Description

Specifications

Specifications

Vinyl
6 in. dissecting mall probe chrome, 2 dissecting teasing needle straight with metal chuck, 4.5 in. dissecting student dissecting scissors, dissecting scalpel, 4.5 in. dissecting student forceps chrome plated, glass dropper, 6 in. ruler
Each
Biology Dissecting Kit
SDS
Documents