Please login to your online account to display your discounted pricing

Dräger™ X-am™ 5000 Multigas Detector

Super-small one-to-five gas detector

$1,866.97 - $2,377.74

Specifications

Alarm Types Audible, visual
Battery Life 12 hr.
Dimensions (L x W x H) 1.2 x 1.9 x 5.1 in. (3.1 x 4.7 x 12.9cm)
IP Rating 67
Temperature (English) Range -4° to +122°F
View More Specs
Products
Catalog Number Mfr. No. Sensor Types Battery Type Price Quantity    

17-985-615

 
dräger™
4543748
No sensors NiMH Each for $1,866.97

17-985-620

 
dräger™
4543751
Carbon Monoxide, Hydrogen Sulfide, LEL, Oxygen NiMH Each for $2,377.74

17-985-622

 
dräger™
4543753
Ex, Hydrogen Sulfide, Oxygen NiMH Each for $2,222.94

17-985-623

 
dräger™
4543754
Ex, Oxygen NiMH Each for $2,256.07
Description & Specifications

Specifications

Alarm Types Audible, visual
Battery Life 12 hr.
Dimensions (L x W x H) 1.2 x 1.9 x 5.1 in. (3.1 x 4.7 x 12.9cm)
IP Rating 67
Temperature (English) Range -4° to +122°F
Temperature (Metric) Range -20° to +50°C
Weight (English) 7.68 oz.
Weight (Metric) 220g

Practical, mobile phone-sized instrument offers maximum security and extremely low operational costs. Reliably measures combustible gases and vapors as well as oxygen and harmful concentrations of other selected gases.

  • Two-button control panel and easy-to-follow menu system allow for intuitive use
  • Large display window makes it possible to view all sensor readings at a glance
  • Integrated rubber protection and shockproof sensors provide impact and vibration resistance; unit also withstands electromagnetic interference
  • High-performance, miniaturized DrägerSensor™ sensors are easy to exchange, upgrade, and calibrate
  • Special calibrations for the catalytic Ex sensor allow more sensitivity to specific combustible gases and vapors
  • Unit can be used with alkaline or NiMH batteries, or it can be fitted with T4 battery capable of in-instrument charging
  • Redundant alarm system includes 90dB multitone audible alarm, 180-degree visual alarm, and a built-in vibration alarm
  • Fresh air, mixed gas, and single gas calibrations can be done directly from menu
  • Functions in a temperature range of -4° to +122°F (-20° to +50°C); relative humidity, 10 to 95%
  • Offers secure gas entry through inlets on both sides of instrument
  • Datalogger can be read out via infrared; stores 1000 hours (6 months) of data for five gases, with recording interval of one minute
  • Optional external pump with 65.6 ft. (20m) hose makes pre-entry measurement of tanks and shafts possible; call for ordering information

Dimensions (L x W x H): 1.2 x 1.9 x 5.1 in. (3.1 x 4.7 x 12.9cm). Weight: approx. 7.8 oz. (220g). Run time: >12 hours (>40 hours in pulsed mode).

IP67 rated for ingress. UL Classified as Intrinsically Safe for Class I, II, Div. 1, Group A, B, C, D, E, F, G hazardous locations, Temp Code T4/T3; ATEX, I M 1 Ex ia I, II 1 G Ex ia IIC T3, I M 2 Ex d ia I, and II 2G Ex d ia IIC T4/T3. CE mark.

Complies with EMC Directive 89/336/EWG, RoHS Directive 2002/95/EC.