Dräger™ X-am™ 3000 Gas Monitor: Plastic Sample Probe
Plastic sample probe for X-am 3000 gas monitors
Manufacturer:  Dräger™ 4511262
Catalog No. 19-038-639
Description
Description
For use with X-am 3000 gas monitors
  • Clear
  • Plastic
  • Length: 12 in.
Specifications
Specifications
Sample Probe
12 in.
Plastic