Dräger™ Short-Term Detector Tubes - Phosphine
Manufacturer:  Dräger™ 8101621
 View more versions of this product
Catalog No. 29-663-371
Description
Description
Specifications
Specifications
Short-Term Gas Detector Tubes
Less than30mg/L
1.5 to 10 min.
10
0° to 40°C
Phosphine 25/a
25 to 10000ppm
1/10
32° to 104°F