missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dräger™ Panorama Nova RA Full-Facepiece

$534.38 - $636.63

Specifications

Certifications/Compliance NIOSH, EN 136
Product Type Full Facepiece
Suspension Type 5-point
Weight (English) 1.2 to 1.4 lb.
Weight (Metric) 0.55 to 0.65 kg
View More Specs

Products 2
Catalog Number Mfr. No. Includes Price Quantity  
Catalog Number Mfr. No. Includes Price Quantity  
19-815-844
View Documents Promotion Details
Dräger™
R51525
Centrally located filter, neck strap
Each for $534.38
 
19-500-224
View Documents Promotion Details
Dräger™
R51535
Centrally located filter
Each for $636.63
 
Description

Description

Classic, full-facepiece mask features triple sealing surfaces for a close, secure and comfortable fit without added pressure. Universal size fits all face types and eliminates the need for different size facepieces. An adjustable five-point rubber head strap aids in quick donning and doffing, making it ideal for use with SCBAs.

Features:
  • Shock-resistant polycarbonate lens provides a wide field of vision.
  • Elaborate flushing system prevents field of vision from being compromised and reduces fogging
  • Stainless-steel speech diaphragm ensures clear, reliable oral communication
  • Robust synthetic connector has both inhalation and exhalation valves
  • RA-type thread connection conforms to European standard EN 148-1 and accommodates a wide variety of Dräger respiratory filters

Order Info

Weight: 1.2 lb. (0.6kg)
Contact your Fisher Safety Customer Service Representative at 1-800-772-6733 for information about Dräger respiratory filters to fit your application.

Compliance

NIOSH approved
Both facepieces meet EN 136 standards

Specifications

Specifications

NIOSH, EN 136
5-point
0.55 to 0.65 kg
Full Facepiece
1.2 to 1.4 lb.
SDS
Product Certifications