Please login to your online account to display your discounted pricing

Dräger™ Pac™ 3500 Single-Gas Detector

Offers two-year, maintenance-free use

$354.69 - $354.69

Specifications

Alarm Types Audible, visual, vibrating
Depth (English) 1 in.
Depth (Metric) 2.5cm
Weight (English) 3.8 oz.
Weight (Metric) 106g
View More Specs
Includes:  Alligator clip for secure attachment
Products
Catalog Number Mfr. No. Measurement Range Sensor Types Price Quantity    

19-500-222

 
dräger™
4543959
0 to 25% Oxygen Each for $354.69
Description & Specifications

Specifications

Alarm Types Audible, visual, vibrating
Depth (English) 1 in.
Depth (Metric) 2.5cm
Weight (English) 3.8 oz.
Weight (Metric) 106g
Height (English) 2.5 in.
Height (Metric) 6.4cm
Humidity Range 10 to 90%
Includes Alligator clip for secure attachment
IP Rating 65
Pressure Range 700 to 1300hpa
Temperature (English) Range -20° to +120°F
Temperature (Metric) Range -30° to +50°C
Width (English) 3.3 in.
Width (Metric) 8.4cm

Ideal for personal monitoring of industrial oxygen, carbon monoxide, or hydrogen sulfide.

  • Compact, robust and ergonomic design with intuitive two-button operation
  • Hardy sensors can be used in extreme environmental conditions without triggering false alarms
  • Large, language-free backlit display continuously indicates gas concentration while simultaneously displaying the corresponding measurement unit
  • Equipped with a sensor that allows gas intake from both the top and front of the unit to ensure continuous measurement, even if a gas inlet is covered
  • Triple alarms for exceptional safety—clear, multi-toned signal, a bright, flashing 360° LED on top and bottom, and vibration
  • Store up to 60 events, along with dates and times; infrared interface enables data to be loaded onto a PC for analysis (requires connecting cradle or E-Cal System and software, sold separately)
  • When bump mode is activated, the unit automatically reminds users when a bump test or calibration is required
  • Operates in a temperature range from -22° to +122°F (-30° to +50°C); relative humidity, 10 to 90%

Dimensions: 0.8L x 2.5W x 3.3 in.H (2.0 x 6.4 x 8.4cm). Weight: 3.8 oz. (106g).

IP65 rated for ingresss protection. UL Classified as Intrinsically Safe for use in Class I, Division 1, Group A, B, C, D, E, F, G hazardous locations, Temp Code T4; ATEX, I/II 1/2 G EEx ia I/IIC, T4. CE mark.

Complies with EMC Directive 89/336/EEC.