Please login to your online account to display your discounted pricing
Dräger™ Installed XXS Dräger™Sensor™ Sensors for X-am™ 5000 Multigas Monitors
$530.33 - $703.78
Specifications
Temperature (English) Range -4° to +122°F
Temperature (Metric) Range -20° to +50°C
Warranty 1 Year
View More Specs
Products
Catalog Number Mfr. No. Measurement Range Resolution Sensor Types Price Quantity    
17-985-639
dräger™
4543768
0 to 300 1 Ammonia (XXS) Each for $530.33
17-985-640
dräger™
4543769
0 to 5% 0.1% Carbon Dioxide (XXS) Each for $703.78
17-985-644
dräger™
4543773
0 to 200 0.5 Organic Vapor (XXS) Each for $530.33
17-980-823
dräger™
4543774
Each for $673.10
Description & Specifications
Specifications
Temperature (English) Range -4° to +122°F
Temperature (Metric) Range -20° to +50°C
Warranty 1 Year
Use to customize your X-am 5000 instrument at time of purchase.