Dräger™ CMS Chips: Petroleum Hydrocarbons
Suitable for 10 measurements of one gas
Manufacturer:  Dräger™ 6406270
 View more versions of this product
Catalog No. 29-801-254
Description
Description
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Specifications
Safety
1 to 30mg/L
30 to 110 sec.
32° to 104°F
700 to 1100hpa
100 to 3000
Petroleum Hydrocarbons CMS™ chip
0° to 40°C
Documents