Dräger™ CMS Chips: Phosphine
Suitable for 10 measurements of one gas.
Manufacturer:  Dräger™ 6406500
 View more versions of this product
Catalog No. 29-801-257
Description
Description
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Specifications
Safety
1 to 30mg/L
20 to 200 sec.
32° to 104°F
700 to 1100hpa
200 to 5000
Phosphine CMS™ chip
0° to 40°C
Documents