Dräger™ CMS Chips: Phosphine
Suitable for 10 measurements of one gas.
Manufacturer:  Dräger™ 6406410
 View more versions of this product
Catalog No. 29-801-255
Description
Description
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Specifications
700 to 1100hpa
1 to 25
Phosphine CMS™ chip
0° to 40°C
1 to 30mg/L
50 to 600 sec.
32° to 104°F