Dräger™ CMS Chips: Nitrous Gases
Suitable for 10 measurements of one gas
Manufacturer:  Dräger™ 6406240
 View more versions of this product
Catalog No. 29-801-251
Description
Description
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Specifications
700 to 1100hpa
10 to 200
Nitrous Gases CMS™ chip
0° to 40°C
1 to 30mg/L
20 to 100 sec.
32° to 104°F