Dräger™ CMS Chips: Petroleum Hydrocarbons
Suitable for 10 measurements of one gas
Manufacturer:  Dräger™ 6406200
 View more versions of this product
Catalog No. 29-801-253
Description
Description
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Specifications
700 to 1100hpa
20 to 500
Petroleum Hydrocarbons CMS™ chip
0° to 40°C
1 to 30mg/L
150 to 330 sec.
32° to 104°F